Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Chemerin Monoclonal Antibody (372402), Invitrogen™
Rat Monoclonal Antibody
Supplier: Thermo Scientific MA524115
Description
Reconstitute at 0.5 mg/mL in sterile PBS. Endoxin level is <0.10 EU per 1 μg of the antibody by the LAL method.
Chemerin Monoclonal antibody specifically detects Chemerin in Mouse samples. It is validated for Neutralization, Western Blot.Specifications
Chemerin | |
Monoclonal | |
0.5 mg/mL | |
PBS with 5% trehalose and No Preservative | |
Q9DD06 | |
Rarres2 | |
E. coli-derived recombinant mouse Chemerin Thr17-Ser156 | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG2a |
Neutralization, Western Blot | |
372402 | |
Unconjugated | |
Rarres2 | |
0610007L05Rik; AI303516; Chemerin; HP10433; Rarres2; RAR-responsive protein TIG2; retinoic acid receptor responder (tazarotene induced) 2; retinoic acid receptor responder 2; retinoic acid receptor responder protein 2; Tazarotene-induced gene 2 protein; TIG2; TIG-2; TIG2HP10433 | |
Rat | |
Protein A/G | |
RUO | |
71660 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction