Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-CHFR, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP130986

Catalog No. NBP130986


Add to cart

Description

Description

CHFR Polyclonal antibody specifically detects CHFR in Human samples. It is validated for Western Blot.
Specifications

Specifications

CHFR
1.0mg/mL
Western Blot 1:500-1:3000
Affinity Purified
CHFR
Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human CHFR. The exact sequence is proprietary.
73 kDa
100 ul
Lipid and Metabolism, Mitotic Regulators, Zinc Finger
Primary
55743
Human
Western Blot
Unconjugated
PBS (pH 7.0) and 40% Glycerol with 0.01% Thimerosal
checkpoint with forkhead and ring finger domains, Checkpoint with forkhead and RING finger domains protein, E3 ubiquitin-protein ligase CHFR, EC 6.3.2.-, FLJ10796, FLJ33629, RING finger protein 196, RNF196RNF116
Rabbit
IgG
Antigen Affinity-purified
RUO
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Polyclonal
Tested in human. Predicted cross-reactivity based on sequence identity: Xenopus laevis (100%) and Xenopus tropicalis (100%). Not yet tested in any other species.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only