Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ChGn (B01P), Mouse anti-Human, Purified MaxPab™ Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a full-length human ChGn protein.

Manufacturer:  Abnova Corporation H00055790B01P

Catalog No. 89-002-850


Add to cart

Description

Description

Sequence: MMMVRRGLLAWISRVVVLLVLLCCAISVLYMLACTPKGDEEQLALPRANSPTGKEGYQAVLQEWEEQHRNYVSSLKRQIAQLKEELQERSEQLRNGQYQASDAAGLGLDRSPPEKTQADLLAFLHSQVDKAEVNAGVKLATEYAAVPFDSFTLQKVYQLETGLTRHPEEKPVRKDKRDELVEAIESALETLNNPAENSPNHRPYTASDFIEGIYRTERDKGTLYELTFKGDHKHEFKRLILFRPFGPIMKVENEKLNMANTLINVIVPLAKRVDKFRQFMQNFRPADEVFRCVPLSP
Specifications

Specifications

ChGn
Unconjugated
In 1X PBS, pH 7.4
BC060772
CSGalNAcT-1|ChGn|FLJ11264|FLJ13760|beta4GalNAcT
Mouse
50 ug
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Polyclonal
Human
Western Blot
Mouse polyclonal antibody raised against a full-length human ChGn protein.
CSGALNACT1
Purified
CSGALNACT1
ChGn (AAH60772, 1 a.a. ~ 297 a.a) full-length human protein.
RUO
Primary
55790
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit