Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CHIC2 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA523975

Catalog No. PIPA523975


Add to cart

Description

Description

CHIC2 is a member of the CHIC family of proteins.This protein contains a cysteine-rich hydrophobic (CHIC) motif, and is localized to vesicular structures and the plasma membrane.
Specifications

Specifications

CHIC2
Polyclonal
PBS with 0.09% sodium azide
Q9UKJ5, Q9D9G3
BTL, BRX-like translocated in leukemia, cystein-rich hydrophobic domain 2, cysteine-rich hydrophobic domain 2 protein
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
26511, 74277
Western Blot
Unconjugated
CHIC2
Liquid
CHIC2
KLH conjugated synthetic peptide between 138-165 amino acids from the C-terminal region of human CHIC2
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit