Learn More
Seracare Life Sciences IncSupplier Diversity Partner ANTI-CHICKEN IGG (H+L) AB  DFS Item
Manufacturer:  Seracare Life Sciences IncSupplier Diversity Partner 52100205
Affinity purified polyclonal antibody to Chicken IgG, both heavy and light chains (whole IgG) made in Goat. Product is in lyophilized form. Each lot is tested to assure specificity and lot-to-lot consistency using KPL's in-house ELISA assay.
Catalog No. 50-668-46