Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chk2 Mouse, Unlabeled, Clone: 19, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 611571

 View more versions of this product

Catalog No. BDB611571


Add to cart

Description

Description

The cell cycle is regulated by multiple checkpoints that determine cell fate. Such checkpoints ensure that DNA replication and chromosomal segregation are completed with high fidelity. When DNA is damaged, specific kinases and phosphatases, key components of cell cycle checkpoints, function to arrest the cell cycle and provide the necessary time for DNA repair. For example, DNA damage induces arrest of the cell cycle at the G2 checkpoint via Wee1-mediated inhibitory phosphorylation of the kinase Cdc2. Inhibition is relieved by the Cdc25C
phosphatase via Cdc2 dephosphorylation. In turn, the Chk1 and Chk2 protein kinases phosphorylate and inhibit Cdc25C, thus preventing activation of the Cdc2-cyclin B complex and entry into mitosis. Chk2, the human homolog of S. cerevisiae Rad53 and S. pombe Cds1, contains a C-terminal kinase domain, an N-terminal regulatory region that is rich in TQ and SQ pairs, and a forked head-associated domain (FHA) which is found in other cell cycle kinases. Chk2 is expressed during late G1 to M phase and is found in the nucleoplasm. Thus, Chk1
and Chk2 activity may be an important checkpoint that inhibits entry into mitosis in response to DNA damage.

Host Species: Mouse
Clone: 19
Isotype: IgG1
Species Reactivity: Mouse
Immunogen: Mouse Chk2 aa. 31-234
Formula Weight [Chemical]: 60kDa

Immunofluorescence, Western Blotting

Specifications

Specifications

Chk2
19
Unconjugated
Affinity Purified
Mouse Chk2 aa. 31-234
150μg
Cell Biology
Primary
Human, Murine, Rat
Western Blot
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Murine
IgG1
RUO
Store undiluted at -20°C.
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only.