Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Chk2, Polyclonal, Novus Biologicals™

Manufacturer:  Novus Biologicals NBP176780

Catalog No. NBP176780

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

Chk2
1 mg/mL
Western Blot 0.5-1 ug/ml, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:10-1:2000
O96017
CHK2 checkpoint homolog (S. pombe), EC 2.7.11, EC 2.7.11.1, S.pombe) homolog
Rabbit
IgG
Immunogen affinity purified
RUO
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Polyclonal
Human, Mouse, Rat
ELISA, Immunocytochemistry, Immunofluorescence, Western Blot
Unconjugated
PBS with 0.02% Sodium Azide
Affinity Purified
CHEK2
Antibody was raised against a synthetic peptide corresponding to amino acids near the amino terminus of human Chk2. The immunogen is located within the first 50 amino acids of Chk2.
60 kDa
0.1 mg
Apoptosis, Breast Cancer, Cancer, Cell Cycle and Replication, Checkpoint signaling, Core ESC Like Genes, DNA Double Strand Break Repair, DNA Repair, Stem Cell Markers
Primary
11200
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only