Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chlamydia trachomatis, Biotin, Polyclonal, Invitrogen™

Catalog No. PIPA128936
Change view
Click to view available options
Quantity:
1 mL
Catalog No. Quantity
PIPA128936 1 mL
1 options

Catalog No. PIPA128936

Supplier: Thermo Scientific PA128936

Goat Polyclonal Antibody

Chlamydia trachomatis is an intracellular organism. Chlamydia usually infects the cervix and fallopian tubes of women and the urethra of men. Chlamydial infections are believed to be one of the most common of all STDs. It is generally thought that in a population of 15 million, there are up to 300,000 cases of chlamydia each year. Thus, there are many undiagnosed cases of chlamydia in the community. It has been estimated that the true prevalence of chlamydia in the sexually active population may be in the order of 5% to 10%. Chlamydia is one of the leading causes of blindness in underdeveloped countries.

PA1-28936 detects Chlamydia trachomatis from bacteria samples.PA1-28936 has been successfully used in immunofluorescence applications.The PA1-28936 immunogen is: Purified MOMP from strain L2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Chlamydia trachomatis
Applications Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Concentration 4.5mg/mL
Conjugate Biotin
Gene Alias C. trachomatis
Host Species Goat
Immunogen Purified MOMP from strain L2.
Purification Method IgG fraction
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.