Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chlamydia trachomatis, FITC, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA129015

Catalog No. PIPA129015

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Chlamydia trachomatis is an intracellular organism. Chlamydia usually infects the cervix and fallopian tubes of women and the urethra of men. Chlamydial infections are believed to be one of the most common of all STDs. It is generally thought that in a population of 15 million, there are up to 300,000 cases of chlamydia each year. Thus, there are many undiagnosed cases of chlamydia in the community. It has been estimated that the true prevalence of chlamydia in the sexually active population may be in the order of 5% to 10%. Chlamydia is one of the leading causes of blindness in underdeveloped countries.

PA1-29015 detects Chlamydia trachomatis (FITC ) from bacteria samples.PA1-29015 has been successfully used in immunofluorescence applications.The PA1-29015 immunogen is: L2 and other serovar groups.
Specifications

Specifications

Chlamydia trachomatis
Polyclonal
FITC
Purified
Rabbit
IgG
100μL
Store at -20°C
Primary
Bacteria
Immunocytochemistry, Immunofluorescence
4.5mg/mL
PBS with 10mg/mL BSA and 0.1% sodium azide; pH 7.2
C. trachomatis
L2 and other serovar groups.
IgG fraction
RUO
Antibody
Polyclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit