Learn More
Chlamydia trachomatis LPS Mouse anti-Bacteria, Clone: CL21-331.1, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA110678
Catalog No. 01-671-909
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
MA1-10678 detects Chlamydia trachomatis LPS from C. trachomatis serovars A, B, Ba, C, D, E, F, G, H, I, J, K, L1, L2, and L3 samples. MA1-10678 may be used in immunoassays to detect C. trachomatis. The MA1-10678 immunogen is C. trachomatis elementary bodies.
Chlamydia trachomatis is an intracellular organism. Chlamydia usually infects the cervix and fallopian tubes of women and the urethra of men. Chlamydial infections are believed to be one of the most common of all STDs. It is generally thought that in a population of 15 million, there are up to 300,000 cases of chlamydia each year. Thus, there are many undiagnosed cases of chlamydia in the community. It has been estimated that the true prevalence of chlamydia in the sexually active population may be in the order of 5% to 10%. Chlamydia is one of the leading causes of blindness in underdeveloped countries.
Specifications
Specifications
Chlamydia trachomatis LPS
Monoclonal
0.9 mg/mL
PBS with no preservative; pH 7.4
C. trachomatis Lipopolysaccharide; Chlamydia trachomatis Lipopolysaccharide
C. trachomatis elementary bodies.
Purified
RUO
Antibody
Monoclonal
ELISA
CL21-331.1
Unconjugated
Liquid
Mouse
IgG2a
500 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
Bacteria