Learn More
Chlamydia trachomatis MOMP Mouse anti-Bacteria, Clone: CL12-741.24.5.5, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA110668
Catalog No. 01-671-599
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
MA1-10668 detects Chlamydia trachomatis MOMP from C. trachomatis serovars A, B, Ba, C, D, E, F, G, H, I, J, K, L1, L2, and L3 samples. MA1-10668 has been successfully used in ELISA applications. The MA1-10668 immunogen is C. trachomatis elementary bodies.
Chlamydia trachomatis is an intracellular organism. Chlamydia usually infects the cervix and fallopian tubes of women and the urethra of men. Chlamydial infections are believed to be one of the most common of all STDs. It is generally thought that in a population of 15 million, there are up to 300,000 cases of chlamydia each year. Thus, there are many undiagnosed cases of chlamydia in the community. It has been estimated that the true prevalence of chlamydia in the sexually active population may be in the order of 5% to 10%. Chlamydia is one of the leading causes of blindness in underdeveloped countries.
Specifications
Specifications
Chlamydia trachomatis MOMP
Monoclonal
∽1 mg/mL
PBS with no preservative; pH 7.4
C. trachomatis major outer membrane protein; C. trachomatis MOMP
C. trachomatis elementary bodies.
Purified
RUO
Antibody
Monoclonal
ELISA
CL12-741.24.5.5
Unconjugated
Liquid
Mouse
IgG2a
500 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
Bacteria