Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Chlamydia trachomatis MOMP Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Supplier: Thermo Scientific PA17209
Description
PA1-7209 detects chlamydia trachomatis. This antibody does not react with C. psittacii or C. pneumoniae in MIF. PA1-7209 has been successfully used in immunofluorescence procedures. Potential applications for this product are numerous including ELISA, fluorescence microscopy, immunoblotting and immunohistochemistry. The PA1-7209 immunogen is purified MOMP from strain L2.
Chlamydia trachomatis MOMP Polyclonal antibody specifically detects Chlamydia trachomatis MOMP in Bacteria samples. It is validated for Immunocytochemistry.Specifications
Chlamydia trachomatis MOMP | |
Polyclonal | |
Unconjugated | |
C. trachomatis MOMP; Chlamydia MOMP | |
Purified MOMP from strain L2. | |
1 mL | |
Primary | |
Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C | |
Liquid |
Immunocytochemistry | |
4-5 mg/mL | |
PBS with 0.1% sodium azide; pH 7.2 | |
Goat | |
purified | |
RUO | |
Bacteria | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction