Learn More
Cygnus Technologies ANTI-CHO:HRP CONJUGATE, 3G
Supplier:  Cygnus Technologies F551112
Encompass
For use in ELISA. Based on the anti-CHO HCP antibody utilized in CHO HCP ELISA, 3G (F550-1). 12 mL.
Catalog No. NC2243578
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.