Learn More
Cholera Toxin beta Rabbit anti-Bacteria, HRP, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA185293
Catalog No. PA185293
Add to cart
Description
Description
PA1-85293 is specific for the beta subunit of Cholera toxin in bacterial samples. This product has an Enzyme: Protein (Molar Ratio) of 2-3. Rabbit anti cholera toxin beta antibody recognizes the beta subunit of cholera toxin.
Cholera toxin (sometimes abbreviated to CTX, Ctx, or CT) is a protein complex secreted by the bacterium Vibrio cholerae. CTX is the causative agent of cholera infection. The alpha subunit contains two chains, A1 and A2, linked by a disulfide bridge. The alpha subunit activates the adenylate cyclase enzyme in cells of the intestinal mucosa leading to increased levels of intracellular cAMP. The beta subunit of cholera toxin binds to a GM1-ganglioside receptor which is widely accepted to initiate toxin action by triggering uptake and delivery of the toxin alpha subunit into cells. The holotoxin consists of a pentameric ring of beta subunits whose central pore is occupied by the alpha subunit.
Specifications
Specifications
Cholera Toxin beta
Polyclonal
HRP
Liquid
Rabbit
IgG
1 mL
4°C or -20°C if preferred
Primary
Bacteria
ELISA, Western Blot
1 mg/mL
PBS with 1% BSA and 0.002% thimerosal; pH 7.2
beta subunit cholera toxin; CTB; VC1456; CTX
Purified Choleragenoid.
Purified
RUO
Antibody
Polyclonal