anti-Choline Acetyltransferase Ab-4; Polyclonal; Millipore™ (Chemicon™)

Manufacturer: EMD Millipore AB144/DEL

 View more versions of this product

Catalog No. 50-173-059

 N/A
N/A

This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »


    Specifications

    Antigen Choline Acetyltransferase Ab-4
    Format Serum
    Host Species Goat
    Immunogen Human placental enzyme.
    Quantity 50μL
    Regulatory Status RUO
    Research Discipline Neuroscience
    Primary or Secondary Primary
    Monoclonal or Polyclonal Polyclonal
    Conjugate Unlabeled
    Cross Reactivity Guinea Pig,Mouse,Rat

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit