Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ anti-Choline Transporter, Clone: 62-2E8, ™ (Chemicon™)
Supplier: MilliporeSigma™ MAB5514/DEL

Description
Immunocytochemistry, Immunohistochemistry, Western Blotting

Specifications
Choline Transporter | |
Unconjugated | |
Recombinant protein from the C-terminus of human CHT. | |
RUO | |
Primary | |
Ascites |
Monoclonal | |
Mouse | |
100 μL | |
Neuroscience | |
Human, Mouse | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction