Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CHRDL1 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524162

Catalog No. PIPA524162


Add to cart

Description

Description

CHRDL1 is an antagonist of bone morphogenetic protein 4. The encoded protein may play a role in topographic retinotectal projection and in the regulation of retinal angiogenesis in response to hypoxia. Alternatively spliced transcript variants encoding different isoforms have been described.
Specifications

Specifications

CHRDL1
Polyclonal
PBS with 0.09% sodium azide
Q9BU40
CHL, NRLN1, VOPT, dA141H5.1, chordin-like protein 1, neuralin 1, neuralin-1, neurogenesin-1, ventroptin
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
91851
Immunohistochemistry, Western Blot
Unconjugated
CHRDL1
Liquid
CHRDL1
KLH conjugated synthetic peptide between 2-30 amino acids from the N-terminal region of human CHRDL1
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit