Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-CHRFAM7A, Polyclonal, Novus Biologicals™

Goat Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP136946

Catalog No. NBP136946


Add to cart

Description

Description

CHRFAM7A Polyclonal antibody specifically detects CHRFAM7A in Human samples. It is validated for ELISA, Peptide ELISA.
Specifications

Specifications

CHRFAM7A
0.5 mg/mL
ELISA 1:200, Peptide ELISA Detection limit 1:1000
NP_647536.1
CHRNA7, CHRNA7 (cholinergic receptor, nicotinic, alpha 7, exons 5-10) and FAM7A (familywith sequence similarity 7A, exons A-E) fusion, CHRNA7 (cholinergic receptor, nicotinic, alpha polypeptide 7, exons 5-10) andFAM7A (family with sequence similarity 7A, exons A-E) fusion, CHRNA7-DR1alpha 7 neuronal nicotinic acetylcholine receptor-FAM7A hybrid, CHRNA7-FAM7A fusion protein, D-10alpha-7 nicotinic cholinergic receptor subunit, MGC120482, MGC120483
Goat
IgG
0.1 mg
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
This antibody is expected to recognize reported isoform NP_647536.1 only.
ELISA
Unconjugated
0.5 mg/ml Tris (pH 7.3) and 0.5% BSA with 0.02% Sodium Azide
Affinity Purified
CHRFAM7A
Peptide with sequence QKYCIYQHFQFQ corresponding to N-Terminus according to NP_647536.1.
Immunogen affinity purified
RUO
Primary
89832
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only