Learn More
CHRNE Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA124273
Description
PA1-24273 detects CHRNG in Human samples. PA1-24273 has been successfully used in Western Blot procedures. The PA1-24273 immunogen is a synthetic peptide corresponding to a region of human CHRNG. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.
After binding acetylcholine, the ACHR responds by an extensive change in conformation that affects all subunits and leads to opening of an ion-conducting channel across the plasma membrane.Specifications
CHRNE | |
Polyclonal | |
Unconjugated | |
Q04844 | |
CHRNE | |
Synthetic peptide corresponding to a region of human CHRNG. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
1 mg/mL | |
CHRNE | |
ACHRG, MGC133376 | |
Rabbit | |
Protein A/G | |
RUO | |
1145 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.