Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Chromogranin A Mouse anti-Human, Clone: 23A1, Invitrogen™
Mouse Monoclonal Antibody
Supplier: Thermo Scientific LFMA0140
Description
A suggested positive control for this product is Bosc23 cells transfected with myc/His-CGA.
Chromogranin A (CHGA) is a member of the chromogranin/secretogranin family of neuroendocrine secretory proteins. It is found in secretory vesicles of neurons and endocine cells. CHGA is a precursor to three biologically active peptides: vasostatin, pancreastatin, and parastatin. These peptides act as autocine or paracrine negative modulators of the neuroendocrine system. CHGA is an excellent marker for carcinoid tumors, pheo-chromocytomas, paragangliomas, and other neuro-endocrine tumors. Coexpression of chromogranin A and neuron specific enolase (NSE) is common in neuroendocrine neoplasms.Specifications
Chromogranin A | |
Monoclonal | |
Unconjugated | |
P10645 | |
CHGA | |
Recombit human CGA protein purified from E.coli | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG2b |
Western Blot | |
23A1 | |
CHGA | |
CGA | |
Mouse | |
Ammonium sulfate precipitation | |
RUO | |
1113 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title