Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chromogranin A Mouse anti-Human, Unlabeled, Clone: S21-537, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 564562

Catalog No. BDB564562


Add to cart

Description

Description

Chromogranin A (CGA) is a member of the granin family of regulated secretory proteins that are found in secretory granules in endocrine and neuroendocrine cells and released in response to extracellular stimulation. Intracellularly, granins are important for targeting peptide hormones and neurotransmitters by their ability to aggregate in the low pH, high calcium environment of the trans-Golgi network. Extracellularly, peptides formed from proteolytic processing of granins regulate hormone secretion. CGA is a prohormone that can be cleaved into several biologically active peptides, such as pancreastatin, β-granin, vasostatin, catestatin, and parastatin. β- granin is an N-terminal fragment of CGA, while pancreastatin and catestatin are processed from the central region of CGA. Cells of the adrenal medulla, anterior pituitary, cerebral cortex as well as beta cells of the pancreas and a variety of tumor cell lines express CGA. The expression of CGA can be used to monitor the pancreatic differentiation of pluripotent stem cells.

Specifications

Specifications

Chromogranin A
S21-537
Unconjugated
Aqueous buffered solution containing ≤0.09% sodium azide.
Murine
IgG1, κ
0.1mg
Store undiluted at 4°C
Monoclonal
Flow Cytometry
0.5mg/mL
CGA; CgA; CHGA; Chromogranin-A; CMGA; SP-I
Affinity Purified
Human Chromogranin A Recombinant Protein
The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography.
RUO
Primary
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit