Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Chromogranin A Rabbit anti-Human, Mouse, Non-human primate, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer: Invitrogen PA137445
Description
Heat-mediated antigen retrieval is recommended prior to staining, using a 10mM citrate buffer, pH 6.0, for 10 minutes followed by cooling at room temperature for 20 min. Following antigen retrieval, incubate samples with primary antibody for 10 min at room temperature. A suggested positive control is pancreas tissue.
Chromogranin A (CHGA) is a member of the chromogranin/secretogranin family of neuroendocrine secretory proteins. It is found in secretory vesicles of neurons and endocine cells. CHGA is a precursor to three biologically active peptides: vasostatin, pancreastatin, and parastatin. These peptides act as autocine or paracrine negative modulators of the neuroendocrine system. CHGA is an excellent marker for carcinoid tumors, pheo-chromocytomas, paragangliomas, and other neuro-endocrine tumors. Coexpression of chromogranin A and neuron specific enolase (NSE) is common in neuroendocrine neoplasms.Specifications
Chromogranin A | |
Polyclonal | |
PBS with 1% BSA and 0.1% sodium azide | |
P26339, P10645 | |
CGA, SP-I, betagranin N-terminal fragment of chromogranin A, chromogranin-A, parathyroid secretory protein 1, pituitary secretory protein I | |
Rabbit | |
IgG | |
1 mL | |
-20°C | |
Primary | |
1113, 12652 |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
CHGA | |
Liquid | |
CHGA | |
Recombit protein encoding human chromogranin A. | |
Antigen affinity chromatography | |
RUO | |
Antibody | |
Polyclonal | |
Human, Mouse, Non-human primate |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title