Learn More
anti-CHST11, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP232761
Catalog No. NBP232761
Add to cart
Description
Description
CHST11 Polyclonal antibody specifically detects CHST11 in Zebrafish samples. It is validated for Western Blot.
Specifications
Specifications
CHST11
0.5 mg/mL
Western Blot 1:1000
Q7T3S3
C4S-1, C4ST, C4St-1, C4ST1EC 2.8.2.5, carbohydrate (chondroitin 4) sulfotransferase 11, carbohydrate sulfotransferase 11, Chondroitin 4-O-sulfotransferase 1, Chondroitin 4-sulfotransferase 1, DKFZp667A035, FLJ41682, HSA269537, IgH/CHST11 fusion
Rabbit
IgG
Protein A purified
RUO
Primary
50515
Western Blot
Unconjugated
PBS with 0.09% Sodium Azide
Affinity Purified
CHST11
This DANREchst11 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 131-155 amino acids from the Central region of DANREchst11.
40.998 kDa
400 ul
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Polyclonal
Zebrafish