Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CHST2 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524197

Catalog No. PIPA524197

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This antibody is predicted to react with mouse based on sequence homology.

N-acetylglucosamine-6-O-sulfotransferases, such as CHST2, catalyze the transfer of sulfate from 3-prime-phosphoadenosine5-prime-phosphosulfate (PAPS) to position 6 of a non reducingN-acetylglucosamine (GlcNAc) residue.
Specifications

Specifications

CHST2
Polyclonal
PBS with 0.09% sodium azide
Q9Y4C5
C6ST, GST-2, GST2, Gn6ST-1, glcNAc6ST-1, N-acetylglucosamine 6-O-sulfotransferase 1, carbohydrate (chondroitin 6/keratan) sulfotransferase 2, carbohydrate sulfotransferase 2, galactose/N-acetylglucosamine/N-acetylglucosamine 6-O-sulfotransferase 2
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
9435
Flow Cytometry, Western Blot
Unconjugated
CHST2
Liquid
CHST2
KLH conjugated synthetic peptide between 309-335 amino acids from the central region of human CHST2
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit