Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CHST3 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA523963

Catalog No. PIPA523963


Add to cart

Description

Description

This antibody is predicted to react with rat based on sequence homology.

CHST3 is an enzyme which catalyzes the sulfation of chondroitin, a proteoglycan found in the extracellular matrixand most cells which is involved in cell migration and differentiation.
Specifications

Specifications

CHST3
Polyclonal
PBS with 0.09% sodium azide
Q7LGC8, O88199
C6ST, C6ST1, HSD, C6ST-1, GST, carbohydrate sulfotransferase 3, chondroitin 6-O-sulfotransferase 1, galactose/N-acetylglucosamine/N-acetylglucosamine 6-O-sulfotransferase
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
53374, 9469
Immunohistochemistry, Western Blot
Unconjugated
CHST3
Liquid
CHST3
KLH conjugated synthetic peptide between 25-54 amino acids from the N-terminal region of human CHST3
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit