Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CHX10, Polyclonal, Invitrogen™

Catalog No. PIPA131286
Change view
Click to view available options
Quantity:
250 μg
Catalog No. Quantity
PIPA131286 250 μg
1 options

Catalog No. PIPA131286

Supplier: Thermo Scientific PA131286

Sheep Polyclonal Antibody

Human CHX10 is expressed in progenitor cells of the developing neuroretina and in the inner nuclear layer of the mature retina. The strong conservation in vertebrates of the CHX10 sequence, pattern of expression and loss-of-function phenotypes demonstrate

PA1-31286 detects CHX10 from bovine, rat, human, mouse samples.PA1-31286 has been successfully used in Western blot applications.The PA1-31286 immunogen is: Recombinant fragment: EAAAEKPEGERQALPKLDKMEQDERGPDAQAAISQEELRENSIAVLRAKAQEHSTKVLGTVSGPDSLARSTEKPEEEEAMDEDRPAERLSPPQLEDMA, corresponding to C terminal amino acids 264-361 of Human CHX10.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CHX10
Applications Western Blot
Classification Polyclonal
Concentration 1mg/mL
Conjugate Unconjugated
Gene VSX2
Gene Accession No. P58304, Q61412
Gene Alias CHX10, HOX10, MCOP2, MCOPCB3, RET1
Gene Symbols VSX2
Host Species Sheep
Immunogen Recombinant fragment: EAAAEKPEGERQALPKLDKMEQDERGPDAQAAISQEELRENSIAVLRAKAQEHSTKVLGTVSGPDSLARSTEKPEEEEAMDEDRPAERLSPPQLEDMA, corresponding to C terminal amino acids 264-361 of Human CHX10.
Purification Method IgG fraction
Quantity 250 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 338917
Target Species Bovine, Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.