Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IGF2R Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA3850
Description
PA3-850 detects cation-independent mannose-6-receptor(CI-MPR) in human, bovine, mouse and non-human primate cells. PA3-850 has been successfully used in ICC/IF, Dot blot and Western blotting procedures. By Western blot, it detects a 300 kDa protein representing CI-MPR under non-reducing conditions. The PA3-850 immunogen is a 300 kDa cation-independent receptor purified from adult Bovine liver tissue.
IGF2R Polyclonal antibody specifically detects IGF2R in Bovine, Human, Mouse, Non-human Primate samples. It is validated for Dot Blot, Immunocytochemistry, Western Blot.Specifications
IGF2R | |
Polyclonal | |
Unconjugated | |
IGF2R | |
300 kDa mannose 6-phosphate receptor; AI661837; cation-independent mannose 6-phosphate receptor; cation-independent mannose 6-phosphate receptor protein; cation-independent mannose-6-phosphate receptor; cation-independent MPR; CD222; CI Man-6-P receptor; CI-M6PR; CIMPR; CI-MPR; Igf2r; IGF-II receptor; IGF-II/CI-MPR; Igfr2; insulin like growth factor 2 receptor; insulin-like growth factor 2 receptor; Insulin-like growth factor II receptor; insulin-like growth factor receptor type 2; M6P; M6P/IGF2 receptor; M6P/IGF2R; M6PR; M6P-R; mannose-6-phosphate receptor, cation independent; mannose-6-phosphate/insulin-like growth factor II receptor; MPR 300; MPR1; MPR300; MPRI; similar to mannose-6-phosphate/insulin-like growth factor II receptor | |
Rabbit | |
100 μL | |
Primary | |
Bovine, Human, Mouse, Non-human Primate | |
Antibody | |
IgG |
Dot Blot, Immunocytochemistry, Western Blot | |
Conc. Not Determined | |
whole serum with 0.05% sodium azide | |
P08169, P11717, Q07113 | |
IGF2R | |
300 kDa cation-independent receptor purified from adult bovine liver tissue. | |
RUO | |
16004, 281849, 3482 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction