anti-cIAP-1/HIAP-2, Polyclonal, Novus Biologicals™

Manufacturer: Novus BiologicalsSupplier Diversity Partner 29690002

 View more versions of this product

Catalog No. 29-690-002

  Check Availability
Add to Cart

Disclaimers:

For Research Use Only


    Specifications

    Formulation 20mM Potassium phosphate, 150mM Sodium Chloride, pH 7.0
    Gene Accession No. Q13490
    Host Species Rabbit
    Quantity 0.05mg
    Primary or Secondary Primary
    Gene ID (Entrez) 329
    Test Specificity This product is specific for Human BIRC2.
    Conjugate Unlabeled
    Antigen cIAP-1/HIAP-2
    Format Affinity Purified
    Applications ELISA,Immunohistochemistry,Immunohistochemistry (Paraffin),Western Blotting
    Immunogen This antibody is specific for the N Terminus Region of the target protein.
    Regulatory Status RUO
    Monoclonal or Polyclonal Polyclonal
    Cross Reactivity Human

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit