Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CIDEC Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA14316

Catalog No. PA14316


Add to cart

Description

Description

PA1-4316 detects the human CIDE-3 protein in human samples. Expected to recognize mouse samples due to 93% sequence homology. PA1-4316 has been successfully used in Western blot procedures. By Western blot this antibody detects a ~27 kDa protein representing human CIDE-3 in HepG2 cell lysate. The PA1-4316 immunogen is a synthetic peptide corresponding to residues L(10) L Y P K S L S R H V S V R T S(25) of human CIDE-3. This antibody shows cross-reactivity to FSP27, which is the mouse homolog of CIDE-3, with a sequence identity of 93%. Maintain at 4°C for 3 months or at -20°C for longer periods. Avoid repeated freeze-thaw cycles.

DNA fragmentation is one of the critical steps in apoptosis, which is induced by DNA fragmentation factor (DFF). DFF is composed of two subunits, a 40 kDa caspase-activated nuclease (DFF40) and a 45 kDa inhibitor (DFF45). Recently a novel family of cell-death-inducing DFF45-like effectors (CIDEs) has been identified. Human CIDE-3 is a novel member of CIDEs. There are 2 transcripts, CIDE-3 and CIDE-3alpha, were present in HepG2 and A375 cells. Consistent with its chromosome localization at 3p25, a region associated with high frequency loss of heterozygosity in many tumours, CIDE-3 may play an important role in prevention of tumorigenesis.  
Specifications

Specifications

CIDEC
Polyclonal
Unconjugated
CIDEC
Liquid
CIDEC
Synthetic peptide corresponding to residues L(10) L Y P K S L S R H V S V R T S(25) of human CIDE-3.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Western Blot
1 mg/mL
PBS with 0.02% sodium azide
Q96AQ7, P56198
Cell death activator CIDE-3, Cell death-inducing DFFA-like effector protein C, Fat-specific protein FSP27 homolog
Rabbit
IgG
100 μg
Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Primary
14311, 63924
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit