Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CIDEB Rabbit anti-Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA130544

Catalog No. PIPA130544


Add to cart

Description

Description

PA1-30544 detects CIDE-B from mouse samples. PA1-30544 has been successfully used in Western blot applications. The PA1-30544 immunogen is: Synthetic peptide corresponding to amino acids 204-219 of mouse CIDE-B

Specifications

Specifications

CIDEB
Polyclonal
Unconjugated
Cideb
Liquid
CIDEB
Synthetic peptide corresponding to amino acids 204-219 of mouse CIDE-B
Affinity chromatography
RUO
Antibody
Polyclonal
Mouse
Western Blot
1.0 mg/mL
PBS with 0.02% sodium azide
O70303
1110030C18Rik, AI790179, CID
Rabbit
IgG
100 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
12684
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit