Learn More
cell death-inducing DFFA-like effector a, Mouse, Clone: 4B9, Abnova™
Mouse monoclonal antibody raised against a full-length recombinant CIDEA.
Supplier: Abnova Corporation H00001149M01
Description
This gene encodes the homolog of the mouse protein Cidea that has been shown to activate apoptosis. This activation of apoptosis is inhibited by the DNA fragmentation factor DFF45 but not by caspase inhibitors. Mice that lack functional Cidea have higher metabolic rates, higher lipolysis in brown adipose tissue and higher core body temperatures when subjected to cold. These mice are also resistant to diet-induced obesity and diabetes. This suggests that in mice this gene product plays a role in thermogenesis and lipolysis. Two alternative transcripts encoding different isoforms have been identified. [provided by RefSeq
Sequence: MRGDRASGGPGNHNGSWAREGPRLGPSWKRGLWSPRGGPNRPAEPSRPLTFMGSQTKRVLFTPLMHPARPFRVSNHDRSSRRGVMASSLQELISKTLDALVIATGLVTLVLEEDGTVVDTEEFFQTLGDNTHFMILEKGQKWMPGSQHVPTCSPPKRSGIARVTFDLYRLNPKDFIGCLNVKATMYEMYSVSYDIRCTGLKGLLRSLLRFLSYSAQVTGQFLIYLGTYMLRVLDDKEERPSLRSQAKGRFTCGSpecifications
cell death-inducing DFFA-like effector a | |
Monoclonal | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
BC031896 | |
CIDEA | |
CIDEA (AAH31896, 1 a.a. ∼ 253 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Apoptosis | |
1149 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG2a κ |
ELISA, Immunoprecipitation, Western Blot | |
4B9 | |
Mouse monoclonal antibody raised against a full length recombinant CIDEA. | |
CIDEA | |
CIDE-A | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Human | |
Antibody |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.