Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TSPYL2 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA522808

Catalog No. PIPA522808


Add to cart

Description

Description

DENTT (Differentially Expressed Nucleolar TGF-Beta1 Target) protein, is expressed in several normal tissues with the highest level of expression in brain. DENTT is involved in responses mediated by TGF-Beta1 and has distinct domains for direction to different subnuclear locations.
Specifications

Specifications

TSPYL2
Polyclonal
Unconjugated
TSPYL2
Liquid
TSPYL2
Human DENTT protein
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Western Blot
1 mg/mL
PBS with 0.02% sodium azide
Q9H2G4
RP1-290F12.2, CDA1, CINAP, CTCL, DENTT, HRIHFB2216, NP79, SE204, TSPX, CASK-interacting nucleosome assembly protein, CTCL tumor antigen se20-4, CTCL-associated antigen se20-4, TSPY-like protein 2, cell division autoantigen 1, cutaneous T-cell lymphoma-associated antigen se20-4, cutaneous T-cell lymphoma-associated tumor antigen se20-4, differentially expressed nucleolar TGF-beta1 target, differentially-expressed nucleolar TGF-beta1 target protein, nuclear protein of 79 kDa, testis-specific Y-enc
Rabbit
IgG
100 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
64061
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit