Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CINP, Polyclonal, Invitrogen™

Catalog No. PIPA127737
Change view
Click to view available options
Quantity:
50 μg
Catalog No. Quantity
PIPA127737 50 μg
1 options

Catalog No. PIPA127737

Supplier: Thermo Scientific PA127737

Chicken Polyclonal Antibody

The function of this gene has not been reported. This gene contains a trinucleotide (ctt) repeat length polymorphism; alleles with either one or two repeat units have been identified.

PA1-27737 detects CINP from human samples.PA1-27737 may be successfully used in Western blot applications.The PA1-27640 immunogen is a synthetic peptide corresponding to human full-length cDNA clone CS0DI023YE22 of Placenta of Homo sapiens+F331.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CINP
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Gene CINP
Gene Accession No. Q9BW66
Gene Alias MGC849
Gene Symbols CINP
Host Species Chicken
Immunogen Synthetic peptide corresponding to human Full-length cDNA clone CS0DI023YE22 of Placenta of Homo sapiens+F331.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51550
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Purified
Isotype IgY
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.