Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CIP4 Mouse, Unlabeled, Clone: 21, BD

Mouse Monoclonal Antibody

$232.00

Specifications

Antigen CIP4
Isotype IgG1
Immunogen Human CIP4 aa. 411-501
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
BDB612556
View Documents Promotion Details
BD Biosciences
612556
50μg
Each for $232.00
Add to cart
 
BDB612557
View Documents Promotion Details
BD Biosciences
612557
150μg This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

Rho family members are small GTP binding proteins that serve as molecular switches for a number of biological processes. They cycle between active GTP-bound and inactive GDP-bound states. CDC42 is a Rho family protein that was identified in membranes of human platelets and placenta. It is the homologue of CDC42Sc, which regulates initiation of bud-site assembly in Saccharomyces cerevisiae. Similarly, CDC42 regulates the function of the mammalian actin cytoskeleton, allowing for efficient cytokinesis and cell morphogenesis. CDC42-interacting protein 4 (CIP4) was identified in a yeast-two hybrid screen for proteins that bind CDC42. Another variant of CIP4, CIP4/2, was identified that contains an extra 56 amino acids and has 71% identity with CIP4 (or CIP4/1). CIP4 contains a C-terminal SH3 domain and an N-terminal domain that is homologous to non-catalyitic motifs in the tyrosine kinase Fer. The mRNA expression of CIP4 is highest in skeletal muscle, heart, and placenta. Overexpression of CIP4 in Swiss 3T3 cells reduces the amount of stress fibers and leads to clustering of CIP4 to foci at the dorsal side of the cells. In addition, CIP4 binds the Rho-GTPase activating protein RICH and the cytoskeletal protein WASP. Coexpression of CIP4 and WASP in Cos-7 cells leads to WASP association with microtubules. Thus, CIP4 is involved in various protein-protein interactions associated with cytoskeletal dynamics.

Immunofluorescence, Western Blotting

Specifications

Specifications

CIP4
Human CIP4 aa. 411-501
Unconjugated
Cell Biology
Mouse
Affinity Purified
Monoclonal
Store undiluted at -20°C.
IgG1
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Primary
Western Blot
RUO
21
250μg/mL
SDS
Product Certifications

For Research Use Only.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit