Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CIRH1A Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA526950

Catalog No. PIPA526950


Add to cart

Description

Description

CIRH1A encodes a WD40-repeat-containing protein that is localized to the nucleolus. Mutation of this gene causes North American Indian childhood cirrhosis, a severe intrahepatic cholestasis that results in transient neonatal jaundice, and progresses to periportal fibrosis and cirrhosis in childhood and adolescence.
Specifications

Specifications

CIRH1A
Polyclonal
PBS with 0.09% sodium azide
Q969X6, Q8R2N2
CIRHIN, NAIC, TEX292, UTP4, UTP4, small subunit (SSU) processome component, homolog, cirhin, testis expressed gene 292
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
21771, 84916
Flow Cytometry, Immunohistochemistry, Western Blot
Unconjugated
UTP4
Liquid
UTP4
KLH conjugated synthetic peptide between 116-143 amino acids from the N-terminal region of human CIRH1A
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit