Learn More
CITED1 Mouse anti-Human, Clone: 2H6, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA519360
Catalog No. PIMA519360
Add to cart
Description
Description
Peptide Sequence: MPTTSRPALD VKGGTSPAKE DANQEMSSVA YSNLAVKDRK AVAILHYPGV ASNGTKASGA PTSSSGSPIG SPTTTPPTKP PSFNLHPAPH LLASMQLQKL NSQYQGMAAA TPGQPGEAGP LQNWDFGAQA GGAESLSPSA GAQSPAIIDS DPVDEEVLMS LVVELGLDRA NELPELWLGQ NEFDFTADFP SSC
This gene encodes a member of the CREB-binding protein/p300-interacting transactivator with Asp/Glu-rich C-terminal domain (CITED) family of proteins. The encoded protein, also known as melanocyte-specific gene 1, may function as a transcriptional coactivator and may play a role in pigmentation of melanocytes. Alternatively spliced transcript variants have been described.
Specifications
Specifications
CITED1
Monoclonal
0.5 mg/mL
PBS with no preservative; pH 7.4
Q99966
Cbp/p300-interacting transactivator, with Glu/Asp-rich carboxy-terminal domain, 1, MSG1
Mouse
IgG2a, kappa
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
4435
ELISA, Immunocytochemistry, Immunofluorescence, Western Blot
2H6
Unconjugated
CITED1
Liquid
CITED1
CITED1 (AAH04240, 1 a.a. ∽ 193 a.a) full-length recombit protein with GST tag. MW of the GST tag alone is 26 KDa.
Affinity chromatography
RUO
Antibody
Monoclonal
Human