Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CITED2 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA523683
Description
This antibody is predicted to react with bovine based on sequence homology.
The protein encoded by this gene inhibits transactivation of HIF1A-induced genes by competing with binding of HIF1a to p300-CH1. Mutations in this gene are a cause of cardiac septal defects. Three transcript variants encoding the same protein have been found for this gene.
Specifications
CITED2 | |
Polyclonal | |
CITED2 | |
ASD8, MRG1, P35SRJ, VSD2, MRG-1, MSG-related protein 1, MSG1-related gene 1, cbp/p300-interacting transactivator 2, melanocyte-specific gene 1-related gene 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10370, 17684 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunocytochemistry, Immunofluorescence, Western Blot | |
Unconjugated | |
Q99967, O35740 | |
CITED2 | |
KLH conjugated synthetic peptide between 200-226 amino acids from the C-terminal region of human CITED2 | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction