Learn More
Citrulline Rabbit anti-Chemical, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA186258
Catalog No. PIPA186258
Add to cart
Description
Description
This antibody recognizes conjugated citrulline, but not free citrulline.
Citrulline is an amino acid that is not incorporated into proteins during synthesis but is generated within proteins by the deimination of arginine. Citrulline occurs normally in many proteins, such as Myelin basic protein (MBP) while others, such as fibrin become citrullinated during inflammation or apoptosis.
Specifications
Specifications
Citrulline
Polyclonal
whole serum with 0.1% sodium azide
Conjugated Citrulline
Citrulline-Glutaraldehyde-BSA
100 μL
4°C or -20°C if preferred
Primary
Chemical
ELISA
Unconjugated
Liquid
Rabbit
IgG
RUO
Antibody
Polyclonal