Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MCMBP Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA524664
Description
This antibody is predicted to react with bovine based on sequence homology.
CJ119 is a carefully orchestrated process involving many proteins that assemble at origins of replication. Among these are the 6 proteins of the minichromosome maintece (MCM) complex (e.g., MCM2; MIM 116945), which form a hexamer. Each MCM subunit performs an essential function in initiation and elongation of DNA replication. MCMBP can replace MCM2 in the MCM complex, thus forming an alternative MCM hexamer.Specifications
MCMBP | |
Polyclonal | |
MCMBP | |
C10orf119, MCM-BP, MCM-binding protein, mini-chromosome maintece complex-binding protein, minichromosome maintece complex-binding protein | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
79892 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Flow Cytometry, Immunohistochemistry, Western Blot | |
Unconjugated | |
Q9BTE3 | |
MCMBP | |
KLH conjugated synthetic peptide between 143-171 amino acids from the N-terminal region of human CJ119 | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction