Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C11orf73 Rabbit anti-Hamster, Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524268

Catalog No. PIPA524268

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Required for organization and/or function of the secretory apparatus in Clara cells in lung.
Specifications

Specifications

C11orf73
Polyclonal
PBS with 0.09% sodium azide
Q53FT3
HSPC179, L7RN6, lethal, Chr 7, Rinchik 6, uncharacterized protein C11orf73
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
100762248, 51501
Flow Cytometry, Immunohistochemistry, Western Blot
Unconjugated
HIKESHI
Liquid
Hikeshi
KLH conjugated synthetic peptide between 6-35 amino acids from the N-terminal region of human CK073
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Hamster, Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit