Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CKAP1 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA521400

Catalog No. PIPA521400


Add to cart

Description

Description

PA5-21400 targets CKAP1 in ICC, IF, IHC (P), and WB applications and shows reactivity with Human and mouse samples. The PA5-21400 immunogen is recombit protein fragment contain a sequence corresponding to a region within amino acids 1 and 243 (Q99426) of CKAP1. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

Specifications

Specifications

CKAP1
Polyclonal
Unconjugated
TBCB
Liquid
TBCB
Recombit fragment corresponding to a region within amino acids 1 and 243 of CKAP1 (Uniprot IDNo.Q99426)
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
0.43 mg/mL
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Q99426, Q9D1E6
CG22, CKAP1, CKAPI, cytoskeleton associated protein 1, cytoskeleton-associated protein 1, cytoskeleton-associated protein CKAPI, tubulin-folding cofactor B, tubulin-specific chaperone B
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
1155, 66411
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit