Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-CKAP4/p63, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP126641

Catalog No. NBP126641


Description

Description

CKAP4/p63 Polyclonal antibody specifically detects CKAP4/p63 in Human samples. It is validated for Western Blot, Immunoprecipitation.
Specifications

Specifications

CKAP4/p63
0.2 mg/mL
Western Blot 1:2000-1:10000, Immunoprecipitation 3-5 ug/mg lysate
NP_006816.2
63 kDa membrane protein, CLIMP-63, cytoskeleton-associated protein 4, ERGIC-63, MGC99554, P63, transmembrane protein (63kD), endoplasmic reticulum/Golgi intermediatecompartment, type-II transmembrane protein p63
Rabbit
IgG
0.1 ml
Store at 4C. Do not freeze.
Polyclonal
Human
Western Blot, Immunoprecipitation
Unconjugated
TBS and 0.1% BSA with 0.09% Sodium Azide
Affinity Purified
CKAP4
The immunogen recognized by this antibody maps to a region between residue 375 and 425 of human cytoskeleton-associated protein 4 using the numbering given in entry NP_006816.2 (GeneID 10970).
Immunogen affinity purified
RUO
Primary
10970
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only