Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CKAP4/p63 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
Supplier: Novus Biologicals NBP126643
Description
CKAP4/p63 Polyclonal specifically detects CKAP4/p63 in Human, Mouse, Mammal samples. It is validated for Western Blot, Immunoprecipitation.Specifications
CKAP4/p63 | |
Polyclonal | |
Unconjugated | |
TBS and 0.1% BSA with 0.09% Sodium Azide | |
63 kDa membrane protein, CLIMP-63, cytoskeleton-associated protein 4, ERGIC-63, MGC99554, P63, transmembrane protein (63kD), endoplasmic reticulum/Golgi intermediatecompartment, type-II transmembrane protein p63 | |
Rabbit | |
66 kDa | |
0.1 mL | |
Primary | |
Human, Mouse. Based on 100% sequence identity, this antibody is predicted to react with Chicken, Bovine, Pig, Orangutan, Rhesus Monkey, Gorilla, Chimpanzee, White-tufted-ear marmoset, African elephant, Northern white-cheeked gibbon, and Small-eared galago. | |
Store at 4C. Do not freeze. |
Western Blot, Immunoprecipitation | |
0.2 mg/mL | |
Western Blot 1:2000-1:10000, Immunoprecipitation 3-5 ug/mg lysate | |
Q07065 | |
CKAP4 | |
The immunogen recognized by this antibody maps to a region between residue 552 and 602 of human cytoskeleton-associated protein 4 using the numbering given in entry NP_006816.2 (GeneID 10970). | |
Affinity Purified | |
RUO | |
10970 | |
Human, Mouse, Mammalia | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction