Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CLA-1 Mouse anti-Human, Unlabeled, Clone: 25, BD

Mouse Monoclonal Antibody

$207.00 - $415.00

Specifications

Antigen CLA-1
Isotype IgG1
Immunogen Human CLA-1 aa. 104-294
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
BDB610882
View Documents
BD Biosciences
610882
50μg
Each for $207.00
Add to cart
 
BDB610883
View Documents
BD Biosciences
610883
150μg
Each for $415.00
Add to cart
 
Description

Description

CLA-1 (CD36 and LIMPII Analogous-1) is member of a novel gene family that includes CD36, LIMPII, and SR-BI. CD36 is a cell surface glycoprotein that binds to collagen type I and thrombospondin. LIMPII (lysosomal integral membrane protein II), as its name suggests, is expressed on the membrane of lysosomes. SR-BI (scavenger receptor type B class I) is involved in the selective uptake of cholesterol esters. These proteins include two membrane-anchoring regions, two short cytoplasmic tails, and a large extracellular/luminal domain. CLA-1 mRNA is detected in a wide range of tissues including adrenal glands, liver, and testis. Its expression and similarity with other family members suggest it may play a role in HDL metabolism. However, identification of the CLA-1 receptor on monocytes indicates additional CLA-1 functions in leukocytes.

Immunofluorescence, Western Blotting

Specifications

Specifications

CLA-1
Human CLA-1 aa. 104-294
Unconjugated
Cell Biology
Murine
Affinity Purified
Monoclonal
CD36 and LIMPII Analogous-1
IgG1
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Primary
Western Blot
RUO
25
250μg/mL
Store undiluted at -20°C.
SDS

For Research Use Only.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit