Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CLA, Rat anti-Human, Clone: HECA-452, BV605, BD

Manufacturer:  BD Biosciences 563960

Catalog No. BDB563960


Add to cart

Description

Description

Host Species: Rat
Clone: HECA-452
Isotype: IgM, κ
Species Reactivity: Hu
Immunogen: Lymphocyte-depleted Stromal Preparation of Tonsil

Flow cytometry

Specifications

Specifications

CLA
Affinity Purified
Lymphocyte-depleted Stromal Preparation of Tonsil
RUO
Monoclonal
HECA-452
Rat
100 Tests
Primary
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit