Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CLAC-P Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA14306

Catalog No. PA14306


Add to cart

Description

Description

PA1-4306 detects CLAC-P (NC2-1 region) in human and mouse samples. PA1-4306 has been successfully used in Western blot procedures. By Western blot this antibody detects bands of 50, 70, and 100 kDa representing CLAC-P in formic acid extracts of Alzheimer’s disease brain. PA1-4306 immunizing peptide corresponds to amino acid residues 171-183 of the NC2-1 region of human and mouse CLAC-P.

Antibodies against senile plaque (SP) amyloid obtained a clone 9D2, which labeled amyloid fibrils in SPs and reacted with approximately 50/100 kDa polypeptides in Alzheimer ft.s disease (AD) brains. The 9D2 antigen was identified as CLAC (collagen-like Alzheimer amyloid plaque component), and its precursor as CLAC-P/collagen type XXV. Both secreted and membrane-tethered forms of CLAC-P/collagen type XXV specifically bound to fibrillized Abeta, implicating these proteins in beta-amyloidogenesis and neuronal degeneration in AD.
Specifications

Specifications

CLAC-P
Polyclonal
Unconjugated
COL25A1
Liquid
COL25A1
Synthetic peptide: N(171) H G F L S A D Q QL I K(183)
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Western Blot
1 mg/mL
PBS with 0.02% sodium azide
Q9BXS0, Q99MQ5
CLAC, CLACP
Rabbit
IgG
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
77018, 84570
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit