Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Claspin, Polyclonal, Invitrogen™

Catalog No. PIPA126579
Change view
Click to view available options
Quantity:
50 μL
Catalog No. Quantity
PIPA126579 50 μL
1 options

Catalog No. PIPA126579

Supplier: Thermo Scientific PA126579

Rabbit Polyclonal Antibody

Xenopus claspin is an essential upstream regulator of checkpoint kinase 1 and triggers a checkpoint arrest of the cell cycle in the presence of DNA templates in Xenopus egg extracts. The human gene appears to be the homolog Xenopus claspin and its function has not been determined.

PA1-26579 detects Claspin from human samples.PA1-26579 has been successfully used in Western blot procedures.The PA1-26579 immunogen is a synthetic peptide corresponding to a portion of human human Claspin encoded within exon 15.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Claspin
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1mg/mL
Conjugate Unconjugated
Gene CLSPN
Gene Accession No. Q9HAW4
Gene Alias CLASPIN, MGC131612, MGC131613, MGC131615
Gene Symbols CLSPN
Host Species Rabbit
Immunogen Synthetic peptide corresponding to a portion of human human Claspin encoded within exon 15.
Purification Method Antigen affinity chromatography
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 63967
Target Species Human
Content And Storage Store at 4°C, do not freeze
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.