Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Clathrin Heavy Chain Mouse, Unlabeled, Clone: 23, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 610499

Catalog No. BDB610499


Add to cart

Description

Description

Clathrin is the major protein component in the coat formed around pits and vesicles involved in receptor-mediated endocytosis. Clathrin forms a non-covalently bound triskelion structure composed of three heavy chains (192 kDa each) and three light chains (23-25 kDa each). Each leg of the triskelion structure contains one heavy and one light chain. The three heavy chains forming the triskelion structure are attached at their respective proximal ends like spokes on a wheel. Clathrin heavy chain is composed of a terminal globular domain, a distal segment containing several areas sensitive to enzymatic cleavage, and a proximal segment which contains a light chain binding site. The proximal and distal domains are connected by a joint segment at which there is a sharp bend in the heavy chains of fully-assembled triskelia. Although the calculated molecular weight is 192 kDa, clathrin heavy chain migrates at approximately 180 kDa.

Specifications

Specifications

Clathrin Heavy Chain
23
Unconjugated
Affinity Purified
Aqueous buffered solution containing BSA
50μg
Cell Biology
Primary
Canine, Chicken, Human, Murine, Rat
Western Blot
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Murine
IgG1
RUO
Store undiluted at -20°C.
Monoclonal
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit