Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Claudin 13 Rabbit anti-Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA124420

Catalog No. PIPA124420


Add to cart

Description

Description

PA1-24420 detects Claudin 13 in mouse samples. PA1-24420 has been successfully used in Western Blot procedures. PA1-24420 immunogen corresponds to Polyclonal antibody from rabbits immunized with a synthetic peptide region of mouse Cldn13. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

CLDN13 is a member of claudin family. It is a four-transmembrane protein with WWCC motif, defined by W-X(17-22)-W-X(2)-C-X(8-10)-C.
Specifications

Specifications

Claudin 13
Polyclonal
Unconjugated
Cldn13
Liquid
CLDN13
Synthetic peptide region of human Cldn13.
Protein A
RUO
Antibody
Polyclonal
Mouse
Western Blot
1 mg/mL
PBS with 2% sucrose and 0.09% sodium azide
Q9Z0S4
2500004B14Rik, MGC144291, MGC144292
Rabbit
IgG
100 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
57255
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit