Learn More
anti-Claudin-4, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Manufacturer:  Novus Biologicals NBP215911
Catalog No. NBP215911
Add to cart
Description
Description
Claudin-4 Polyclonal specifically detects Claudin-4 in Human samples. It is validated for Western Blot.
Specifications
Specifications
Claudin-4
1mg/mL
Western Blot 1:500-1:3000
O14493
claudin 4, Clostridium perfringens enterotoxin receptor, Clostridium perfringens enterotoxin receptor 1, CPE-RCPETR, CPE-receptor, hCPE-R, WBSCR8CPERCPETR1claudin-4, Williams-Beuren syndrome chromosomal region 8 protein
Rabbit
IgG
Immunogen affinity purified
RUO
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Polyclonal
Human. Expected to cross react based on sequence identity: Monkey (100%).
Western Blot
Unconjugated
0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal
Affinity Purified
CLDN4
Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human claudin 4. The exact sequence is proprietary.
22 kDa
0.1 ml
Cancer, Extracellular Matrix
Primary
1364
Human